missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-fluorobenzamide, 98%

CAS: 66073-54-9 | C7H5ClFNO | 173.571 g/mol

$56.33 - $178.30

Chemical Identifiers

CAS 66073-54-9
Molecular Formula C7H5ClFNO
Molecular Weight (g/mol) 173.571
MDL Number MFCD00055436
InChI Key KLOZZZNFJYMTNE-UHFFFAOYSA-N
PubChem CID 98564
IUPAC Name 2-chloro-6-fluorobenzamide
SMILES C1=CC(=C(C(=C1)Cl)C(=O)N)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2129406
View Documents
Thermo Scientific Chemicals
B2129406
5 g
Each for $56.33
Only null left
 
AAB2129414
View Documents
Thermo Scientific Chemicals
B2129414
25 g
Each for $178.30
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

66073-54-9
173.571
KLOZZZNFJYMTNE-UHFFFAOYSA-N
2-chloro-6-fluorobenzamide
C7H5ClFNO
MFCD00055436
98564
C1=CC(=C(C(=C1)Cl)C(=O)N)F

Specifications

66073-54-9
C7H5ClFNO
5 g
KLOZZZNFJYMTNE-UHFFFAOYSA-N
2-chloro-6-fluorobenzamide
98564
98%
139°C to 142°C
MFCD00055436
2-fluoro-6-chlorobenzamide, 2-chloro-6-fluoro-benzamide, benzamide, 2-chloro-6-fluoro, acmc-209nth, 6-chloro-2-fluorobenzamide, 2-fluoro-6-chlorobenzoylamino, benzamide,2-chloro-6-fluoro, 2-chloranyl-6-fluoranyl-benzamide
C1=CC(=C(C(=C1)Cl)C(=O)N)F
173.571
173.58
2-Chloro-6-fluorobenzamide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 266-123-9

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only