accessibility menu, dialog, popup

UserName

CAS RN 66073-54-9

IUPAC Name: 2-chloro-6-fluorobenzamide
Synonyms: 2-fluoro-6-chlorobenzamide
Molecular Weight (g/mol): 173.57
Molecular Formula: C7H5ClFNO
InChi Key: KLOZZZNFJYMTNE-UHFFFAOYSA-N
SMILES: NC(=O)C1=C(F)C=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results