missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-fluorobenzamide, 98%

CAS: 66073-54-9 | C7H5ClFNO | 173.571 g/mol

Supplier:  Thermo Scientific Chemicals B2129414

 View more versions of this product

Catalog No. AAB2129414


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Chloro-6-fluorobenzamide
139°C to 142°C
MFCD00055436
2-fluoro-6-chlorobenzamide, 2-chloro-6-fluoro-benzamide, benzamide, 2-chloro-6-fluoro, acmc-209nth, 6-chloro-2-fluorobenzamide, 2-fluoro-6-chlorobenzoylamino, benzamide,2-chloro-6-fluoro, 2-chloranyl-6-fluoranyl-benzamide
C1=CC(=C(C(=C1)Cl)C(=O)N)F
173.571
173.58
66073-54-9
C7H5ClFNO
25 g
KLOZZZNFJYMTNE-UHFFFAOYSA-N
2-chloro-6-fluorobenzamide
98564
98%

Chemical Identifiers

66073-54-9
173.571
KLOZZZNFJYMTNE-UHFFFAOYSA-N
2-chloro-6-fluorobenzamide
C7H5ClFNO
MFCD00055436
98564
C1=CC(=C(C(=C1)Cl)C(=O)N)F
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 266-123-9

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only