missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-nitrobenzotrifluoride, 98%

Catalog No. AAL0662322
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0662322 100 g
AAL0662314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0662322 Supplier Thermo Scientific Chemicals Supplier No. L0662322

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 777-37-7 | C7H3ClF3NO2 | 225.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 777-37-7
Molecular Formula C7H3ClF3NO2
Molecular Weight (g/mol) 225.55
MDL Number MFCD00007296
InChI Key HQROXDLWVGFPDE-UHFFFAOYSA-N
Synonym 2-chloro-5-nitrobenzotrifluoride, 1-chloro-4-nitro-2-trifluoromethyl benzene, 4-chloro-3-trifluoromethyl nitrobenzene, benzene, 1-chloro-4-nitro-2-trifluoromethyl, 3-trifluoromethyl-4-chloronitrobenzene, 2-trifluoromethyl-4-nitrochlorobenzene, 2-chloro-5-nitro-a,a,a-trifluorotoluene, 4-nitro-2-trifluoromethyl chlorobenzene, 1-chloro-2-trifluoromethyl-4-nitrobenzene, 5-nitro-2-chlorobenzotrifluoride
PubChem CID 61213
IUPAC Name 1-chloro-4-nitro-2-(trifluoromethyl)benzene
SMILES [O-][N+](=O)C1=CC=C(Cl)C(=C1)C(F)(F)F

Specifications

Melting Point ∼22°C
Density 1.547
Boiling Point 232°C
Flash Point 98°C (208°F)
Refractive Index 1.508
Quantity 100 g
Beilstein 2216169
Formula Weight 225.55
Percent Purity 98%
Chemical Name or Material 2-Chloro-5-nitrobenzotrifluoride
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H311-H332-H315-H319
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 212-287-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only