missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-5-nitrobenzophenone 98.0+%, TCI America™
$44.73 - $106.25
Chemical Identifiers
| CAS | 34052-37-4 |
|---|---|
| Molecular Formula | C13H8ClNO3 |
| Molecular Weight (g/mol) | 261.661 |
| MDL Number | MFCD00007295 |
| InChI Key | HRPHZUAPQWJPCZ-UHFFFAOYSA-N |
| Synonym | 2-chloro-5-nitrobenzophenone, 2-chloro-5-nitrophenyl phenyl methanone, 2-chloro-5-nitrophenyl-phenylmethanone, 2-chloro-5-nitrophenyl phenyl ketone, acmc-1aifx, methanone, 2-chloro-5-nitrophenyl phenyl, 5-nitro-2-chlorobenzophenone, 2-chloro-5-nitro-benzophenone, hrphzuapqwjpcz-uhfffaoysa |
| PubChem CID | 118591 |
| IUPAC Name | (2-chloro-5-nitrophenyl)-phenylmethanone |
| SMILES | C1=CC=C(C=C1)C(=O)C2=C(C=CC(=C2)[N+](=O)[O-])Cl |
Chemical Identifiers
| 34052-37-4 | |
| 261.661 | |
| HRPHZUAPQWJPCZ-UHFFFAOYSA-N | |
| 118591 | |
| C1=CC=C(C=C1)C(=O)C2=C(C=CC(=C2)[N+](=O)[O-])Cl |
| C13H8ClNO3 | |
| MFCD00007295 | |
| 2-chloro-5-nitrobenzophenone, 2-chloro-5-nitrophenyl phenyl methanone, 2-chloro-5-nitrophenyl-phenylmethanone, 2-chloro-5-nitrophenyl phenyl ketone, acmc-1aifx, methanone, 2-chloro-5-nitrophenyl phenyl, 5-nitro-2-chlorobenzophenone, 2-chloro-5-nitro-benzophenone, hrphzuapqwjpcz-uhfffaoysa | |
| (2-chloro-5-nitrophenyl)-phenylmethanone |
Specifications
| 34052-37-4 | |
| Yellow | |
| MFCD00007295 | |
| 2-chloro-5-nitrobenzophenone, 2-chloro-5-nitrophenyl phenyl methanone, 2-chloro-5-nitrophenyl-phenylmethanone, 2-chloro-5-nitrophenyl phenyl ketone, acmc-1aifx, methanone, 2-chloro-5-nitrophenyl phenyl, 5-nitro-2-chlorobenzophenone, 2-chloro-5-nitro-benzophenone, hrphzuapqwjpcz-uhfffaoysa | |
| C1=CC=C(C=C1)C(=O)C2=C(C=CC(=C2)[N+](=O)[O-])Cl | |
| 261.661 | |
| 261.66 | |
| Crystalline Powder |
| 85°C | |
| C13H8ClNO3 | |
| 5 g | |
| HRPHZUAPQWJPCZ-UHFFFAOYSA-N | |
| (2-chloro-5-nitrophenyl)-phenylmethanone | |
| 118591 | |
| ≥98.0% (GC) | |
| 2-Chloro-5-nitrobenzophenone |
Safety and Handling
TSCA : No