missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-5-nitrobenzophenone 98.0+%, TCI America™
Supplier: TCI America C177625G
Specifications
| 2-Chloro-5-nitrobenzophenone | |
| 85°C | |
| C13H8ClNO3 | |
| 25 g | |
| HRPHZUAPQWJPCZ-UHFFFAOYSA-N | |
| (2-chloro-5-nitrophenyl)-phenylmethanone | |
| 118591 | |
| ≥98.0% (GC) |
| 34052-37-4 | |
| Yellow | |
| MFCD00007295 | |
| 2-chloro-5-nitrobenzophenone, 2-chloro-5-nitrophenyl phenyl methanone, 2-chloro-5-nitrophenyl-phenylmethanone, 2-chloro-5-nitrophenyl phenyl ketone, acmc-1aifx, methanone, 2-chloro-5-nitrophenyl phenyl, 5-nitro-2-chlorobenzophenone, 2-chloro-5-nitro-benzophenone, hrphzuapqwjpcz-uhfffaoysa | |
| C1=CC=C(C=C1)C(=O)C2=C(C=CC(=C2)[N+](=O)[O-])Cl | |
| 261.661 | |
| 261.66 | |
| Crystalline Powder |
Chemical Identifiers
| 34052-37-4 | |
| 261.661 | |
| HRPHZUAPQWJPCZ-UHFFFAOYSA-N | |
| 118591 | |
| C1=CC=C(C=C1)C(=O)C2=C(C=CC(=C2)[N+](=O)[O-])Cl |
| C13H8ClNO3 | |
| MFCD00007295 | |
| 2-chloro-5-nitrobenzophenone, 2-chloro-5-nitrophenyl phenyl methanone, 2-chloro-5-nitrophenyl-phenylmethanone, 2-chloro-5-nitrophenyl phenyl ketone, acmc-1aifx, methanone, 2-chloro-5-nitrophenyl phenyl, 5-nitro-2-chlorobenzophenone, 2-chloro-5-nitro-benzophenone, hrphzuapqwjpcz-uhfffaoysa | |
| (2-chloro-5-nitrophenyl)-phenylmethanone |
Safety and Handling
TSCA : No