accessibility menu, dialog, popup

UserName

CAS RN 34052-37-4

IUPAC Name: (2-chloro-5-nitrophenyl)(phenyl)methanone
Synonyms: (2-chloro-5-nitrophenyl)(phenyl)methanone
Molecular Weight (g/mol): 261.66
Molecular Formula: C13H8ClNO3
InChi Key: HRPHZUAPQWJPCZ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(Cl)C(=C1)C(=O)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)