missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-nitrobenzoic Acid 98.0+%, TCI America™

Catalog No. C0877500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0877500G 500 g
C087725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0877500G Supplier TCI America Supplier No. C0877500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2516-96-3
Molecular Formula C7H4ClNO4
Molecular Weight (g/mol) 201.562
MDL Number MFCD00007294
InChI Key QUEKGYQTRJVEQC-UHFFFAOYSA-N
Synonym 2-chloro-5-nitrobenzoic acid, benzoic acid, 2-chloro-5-nitro, 6-chloro-3-nitrobenzoic acid, 2-chloro-5-nitro-benzoic acid, 2-chloro-5-nitrobenzoicacid, mesalamine impurity m, pubchem13682, acmc-1cb0i, 2-chloro-5-nitrobenzic acid, 5-nitro-2-chlorobenzoic acid
PubChem CID 17287
SMILES C1=CC(=C(C=C1[N+](=O)[O-])C(=O)O)Cl

Specifications

Melting Point 165°C
Color White-Yellow
Quantity 500 g
Formula Weight 201.56
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-5-nitrobenzoic Acid
EINECSNumber (3)-1502
TSCA Yes