missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-methylphenol, 99%

Catalog No. AAL0081614
Change view
Click to view available options
Quantity:
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0081614 25 g
AAL0081622 -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0081614 Supplier Thermo Scientific Chemicals Supplier No. L0081614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 615-74-7 | C7H7ClO | 142.58 g/mol

2-Chloro-5-methylphenol, is used as a pharmaceutical intermediate. It is also used to make other chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-5-methylphenol, is used as a pharmaceutical intermediate. It is also used to make other chemicals.

Solubility
Slightly Soluble (1.2 g/L) (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.

Chemical Identifiers

CAS 615-74-7
Molecular Formula C7H7ClO
Molecular Weight (g/mol) 142.58
MDL Number MFCD00002175
InChI Key SMFHPCZZAAMJJO-UHFFFAOYSA-N
Synonym 6-chloro-m-cresol, phenol, 2-chloro-5-methyl, m-cresol, 6-chloro, 3-methyl-6-chlorophenol, 4-chloro-3-hydroxytoluene, 5-methyl-2-chlorophenol, 6-chloro-3-methylphenol, dsstox_cid_26558, dsstox_rid_81718
PubChem CID 12008
IUPAC Name 2-chloro-5-methylphenol
SMILES CC1=CC=C(Cl)C(O)=C1

Specifications

Melting Point 44°C to 48°C
Density 1.215
Boiling Point 195°C to 197°C
Flash Point 81°C (177°F)
Quantity 25 g
UN Number UN3437
Beilstein 2041487
Solubility Information Slightly Soluble (1.2g/L) (25°C).
Formula Weight 142.59
Percent Purity 99%
Chemical Name or Material 2-Chloro-5-methylphenol
Show More Show Less
Hazard Category H302+H312-H317-H318-H335
Hazard Statement GHS H Statement
H301-H311-H331-H318-H315-H335
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLOROCRESOLS, SOLID
EINECSNumber 210-444-9
RTECSNumber SK4930000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only