missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-methoxyphenol 97.0+%, TCI America™

Catalog No. c27205g
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C27205G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C27205G Supplier TCI America Supplier No. C27205G

Chemical Identifiers

CAS 18113-04-7
Molecular Formula C7H7ClO2
Molecular Weight (g/mol) 158.581
MDL Number MFCD00086345
InChI Key KULSVCANYQIOFD-UHFFFAOYSA-N
Synonym 4-Chloro-3-hydroxyanisole
PubChem CID 140336
IUPAC Name 2-chloro-5-methoxyphenol
SMILES COC1=CC(=C(C=C1)Cl)O

Specifications

Color Yellow
Boiling Point 125°C
Quantity 5 g
Formula Weight 158.58
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloro-5-methoxyphenol