missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-methoxyaniline 98.0+%, TCI America™

Catalog No. C30535G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C30535G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C30535G Supplier TCI America Supplier No. C30535G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2401-24-3
Molecular Formula C7H8ClNO
Molecular Weight (g/mol) 157.597
MDL Number MFCD00047830
InChI Key GBOUQGUQUUPGLO-UHFFFAOYSA-N
Synonym 6-chloro-m-anisidine, 3-amino-4-chloroanisole, benzenamine, 2-chloro-5-methoxy, 2-chloro-5-methoxy-aniline, 6-chloro-m-anisidine hydrochloride, 2-chloro-5-methoxy-phenylamine, 2-chloro-5-methoxybenzenamine hydrochloride, 6-chloro-meta-anisidine hydrochloride, 2-chloro-5-methoxyanilinehydrochloride, timtec-bb sbb008038
PubChem CID 75460
IUPAC Name 2-chloro-5-methoxyaniline
SMILES COC1=CC(=C(C=C1)Cl)N

Specifications

Boiling Point 258°C
Quantity 5 g
UN Number 2810
Formula Weight 157.60
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloro-5-methoxyaniline
EINECSNumber (3)-0908
TSCA No