missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-hydroxybenzoic Acid 97.0+%, TCI America™

Catalog No. C27241G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C27241G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C27241G Supplier TCI America Supplier No. C27241G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 56961-30-9
Molecular Formula C7H5ClO3
Molecular Weight (g/mol) 172.56
MDL Number MFCD04038818
InChI Key UTVCLUZQPSRKMY-UHFFFAOYSA-N
Synonym 2-chloro-5-hydroxybenzoicacid, benzoic acid, 2-chloro-5-hydroxy, 3-carboxy-4-chlorophenol, 2-chloro-5-hydroxybenzenecarboxylic acid, 2-chloro-5-hydroxy-benzoic acid, 2-chloro-5-hydroxy benzoic acid, 2-chloro-5-hydroxybenzene carboxylic acid, pubchem16487, acmc-1au79, ksc269a4t
PubChem CID 458210
IUPAC Name 2-chloro-5-hydroxybenzoic acid
SMILES OC(=O)C1=C(Cl)C=CC(O)=C1

Specifications

Melting Point 180°C
Color Yellow
Quantity 1 g
Formula Weight 172.56
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-5-hydroxybenzoic Acid