missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-5-fluoro-4-pyrimidinone 98.0+%, TCI America™
Spécifications
Spécifications
| Melting Point | 174°C |
| Color | White-Yellow |
| Quantity | 1 g |
| Formula Weight | 148.52 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2-Chloro-5-fluoro-4-pyrimidinone |
Sécurité et manipulation
| TSCA | No |