missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 98.0+%, TCI America™

Catalog No. C31361G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C31361G 1 g
C31365G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C31361G Supplier TCI America Supplier No. C31361G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 444120-94-9
Molecular Formula C11H15BClNO2
Molecular Weight (g/mol) 239.506
MDL Number MFCD03412793
InChI Key IEFBTHROULVUAH-UHFFFAOYSA-N
Synonym 2-chloro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-chloropyridine-5-boronic acid pinacol ester, 2-chloro-5-pyridylboronic acid pinacol ester, 6-chloropyridine-3-boronic acid pinacol ester, 2-chloropyridine-5-boronicacidpinacolester, 2-chloro-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridine, 6-chloro-3-pyridineboronic acid pinacol ester, 6-chloropyridin-3-ylboronic acid pinacol ester, 2-chloro-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-6-chloro-3-pyridyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 2794719
IUPAC Name 2-chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CN=C(C=C2)Cl

Specifications

Melting Point 87°C
Color White
Quantity 1 g
Formula Weight 239.51
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine