missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-(trifluoromethoxy)aniline 97.0+%, TCI America™

Numéro de catalogue. C31421G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
C31421G 1 g
C31425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. C31421G Fournisseur TCI America Code fournisseur C31421G
Il en reste null

Identifiants chimiques

CAS 69695-61-0
Molecular Formula C7H5ClF3NO
Molecular Weight (g/mol) 211.568
MDL Number MFCD01631704
InChI Key YMLWONHDNGICOH-UHFFFAOYSA-N
PubChem CID 2734084
IUPAC Name 2-chloro-4-(trifluoromethoxy)aniline
SMILES C1=CC(=C(C=C1OC(F)(F)F)Cl)N

Spécifications

Color Yellow
Quantity 1 g
Physical Form Clear Liquid at 20°C
Synonym 2-chloro-4-trifluoromethoxy aniline, benzenamine, 2-chloro-4-trifluoromethoxy, 2-chloro-4-trifluoromethoxy phenylamine, benzenamine,2-chloro-4-trifluoromethoxy, 2-chloro-4-trifluoromethoxy-phenylamine, pubchem2941, acmc-209wey, ksc630s5l, 2-chloro4-trifluoromethoxyaniline, 4-trifluoromethoxy-2-chloroaniline
Boiling Point 96°C
Formula Weight 211.57
Percent Purity ≥97.0% (GC,T)
Chemical Name or Material 2-Chloro-4-(trifluoromethoxy)aniline