missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-picoline, 98%

Numéro de catalogue. p-2689977
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Packaging
AC368840250 25 mL Glass bottle
AC368841000 100 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AC368840250 Fournisseur Thermo Scientific Chemicals Code fournisseur 368840250
Il en reste null

CAS: 3678-62-4 | C6H6ClN | 127.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 3678-62-4
Molecular Formula C6H6ClN
Molecular Weight (g/mol) 127.57
MDL Number MFCD00023418
InChI Key MZVSTDHRRYQFGI-UHFFFAOYSA-N
Synonym 2-chloro-4-picoline, pyridine, 2-chloro-4-methyl, 2-chloro-4-methyl-pyridine, 2-chloro-4-methyl pyridine, 2-chloro-gamma-picoline, 4-picoline, 2-chloro, zlchem 438, pubchem1192, acmc-209iop
PubChem CID 77248
IUPAC Name 2-chloro-4-methylpyridine
SMILES CC1=CC(=NC=C1)Cl

Spécifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1510g/mL
Boiling Point 194°C to 195°C
Flash Point 89°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5280 to 1.5300 (20°C, 589nm)
Quantity 25 mL
Beilstein 20,241
Specific Gravity 1.151
Formula Weight 127.57
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Chloro-4-picoline
Afficher plus de résultats Afficher moins de résultats
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 222-951-2

RUO – Research Use Only