missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-4-nitrobenzamide 98.0+%, TCI America™
Supplier: TCI America C282925G
Specifications
| 2-Chloro-4-nitrobenzamide | |
| 171°C | |
| MFCD00017119 | |
| aklomide, aklomix, clomide, alkomide, benzamide, 2-chloro-4-nitro, component of aklomix, component of novastat-w, unii-b0e341ra20, 2-chloro-4-nitro benzamide, dsstox_cid_21405 | |
| C1=CC(=C(C=C1[N+](=O)[O-])Cl)C(=O)N | |
| 200.578 | |
| 200.58 | |
| Crystalline Powder |
| 3011-89-0 | |
| C7H5ClN2O3 | |
| 25 g | |
| GFGSZUNNBQXGMK-UHFFFAOYSA-N | |
| 2-chloro-4-nitrobenzamide | |
| 2075 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 3011-89-0 | |
| 200.578 | |
| GFGSZUNNBQXGMK-UHFFFAOYSA-N | |
| 2075 | |
| C1=CC(=C(C=C1[N+](=O)[O-])Cl)C(=O)N |
| C7H5ClN2O3 | |
| MFCD00017119 | |
| aklomide, aklomix, clomide, alkomide, benzamide, 2-chloro-4-nitro, component of aklomix, component of novastat-w, unii-b0e341ra20, 2-chloro-4-nitro benzamide, dsstox_cid_21405 | |
| 2-chloro-4-nitrobenzamide |
Safety and Handling
TSCA : No