missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-nitrobenzamide 98.0+%, TCI America™

Catalog No. p-7125669
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C28295G 5 g
C282925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C28295G Supplier TCI America Supplier No. C28295G

Chemical Identifiers

CAS 3011-89-0
Molecular Formula C7H5ClN2O3
Molecular Weight (g/mol) 200.578
MDL Number MFCD00017119
InChI Key GFGSZUNNBQXGMK-UHFFFAOYSA-N
Synonym aklomide, aklomix, clomide, alkomide, benzamide, 2-chloro-4-nitro, component of aklomix, component of novastat-w, unii-b0e341ra20, 2-chloro-4-nitro benzamide, dsstox_cid_21405
PubChem CID 2075
IUPAC Name 2-chloro-4-nitrobenzamide
SMILES C1=CC(=C(C=C1[N+](=O)[O-])Cl)C(=O)N

Specifications

Melting Point 171°C
Quantity 5 g
Formula Weight 200.58
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-4-nitrobenzamide