accessibility menu, dialog, popup

UserName

H2NOClNOO

CAS RN 3011-89-0

IUPAC Name: 2-chloro-4-nitrobenzamide
Synonyms: 2-chloro-4-nitrobenzamide
Molecular Weight (g/mol): 200.58
Molecular Formula: C7H5ClN2O3
InChi Key: GFGSZUNNBQXGMK-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC=C(C=C1Cl)[N+]([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)