missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-nitroaniline 98.0+%, TCI America™

Catalog No. C021425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C021425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C021425G Supplier TCI America Supplier No. C021425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 121-87-9
Molecular Formula C6H5ClN2O2
Molecular Weight (g/mol) 172.568
MDL Number MFCD00007665
InChI Key LOCWBQIWHWIRGN-UHFFFAOYSA-N
Synonym ocpna, 2-chloro-4-nitrobenzenamine, o-chloro-p-nitroaniline, 4-nitro-2-chloroaniline, benzenamine, 2-chloro-4-nitro, 1-amino-2-chloro-4-nitrobenzene, aniline, 2-chloro-4-nitro, unii-xsa3zx337b, 2-chloro-4-nitro-phenylamine, ortho-chloro-para-nitroaniline
PubChem CID 8492
IUPAC Name 2-chloro-4-nitroaniline
SMILES C1=CC(=C(C=C1[N+](=O)[O-])Cl)N

Specifications

Melting Point 108°C
Color Yellow
Boiling Point 200°C
Quantity 25 g
UN Number 2237
Formula Weight 172.57
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-4-nitroaniline
EINECSNumber (3)-0407
RTECSNumber BX1400000
TSCA Yes