missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-methylpyrimidine 98.0+%, TCI America™

Catalog No. C26315G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C26315G 5 g
C263125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C26315G Supplier TCI America Supplier No. C26315G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13036-57-2
Molecular Formula C5H5ClN2
Molecular Weight (g/mol) 128.559
MDL Number MFCD00054434
InChI Key BHAKRVSCGILCEW-UHFFFAOYSA-N
Synonym 2-chloro-4-methyl-pyrimidine, pyrimidine, 2-chloro-4-methyl, 2-chloro-6-methylpyrimidine, pubchem6906, chloro-6 methylpyrimidine, 4-methyl-2-chloropyrimidine, 2-chloro-4-methyl pyrimidine, 4-methyl-2-chloro-pyrimidine, ksc493s3l, 4-methyl-2-pyrimidinylchloride
PubChem CID 11629607
IUPAC Name 2-chloro-4-methylpyrimidine
SMILES CC1=NC(=NC=C1)Cl

Specifications

Melting Point 49°C
Color White-Yellow
Boiling Point 94°C
Quantity 5 g
Formula Weight 128.56
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-4-methylpyrimidine