missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-methylpyridine 98.0+%, TCI America™

Numéro de catalogue. C19925G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
C19925G 5 g
C199225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. C19925G Fournisseur TCI America Code fournisseur C19925G
Il en reste null

Identifiants chimiques

CAS 3678-62-4
Molecular Formula C6H6ClN
Molecular Weight (g/mol) 127.571
InChI Key MZVSTDHRRYQFGI-UHFFFAOYSA-N
Synonym 2-chloro-4-picoline, pyridine, 2-chloro-4-methyl, 2-chloro-4-methyl-pyridine, 2-chloro-4-methyl pyridine, 2-chloro-gamma-picoline, 4-picoline, 2-chloro, zlchem 438, pubchem1192, acmc-209iop
PubChem CID 77248
IUPAC Name 2-chloro-4-methylpyridine
SMILES CC1=CC(=NC=C1)Cl

Spécifications

Color Yellow
Boiling Point 99°C
Quantity 5 g
Formula Weight 127.57
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloro-4-methylpyridine