missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-methylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. C30491G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C30491G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C30491G Supplier TCI America Supplier No. C30491G

Specifications

Chemical Name or Material 2-Chloro-4-methylphenylboronic Acid (contains varying amounts of Anhydride)
Melting Point 202°C
CAS 145349-62-8
Color White
Quantity 1 g
Physical Form Crystal-Powder at 20°C
Molecular Formula C7H8BClO2
MDL Number MFCD03411936
Synonym 2-chloro-4-methylphenyl boronic acid, 2-chloro-4-methylbenzeneboronic acid, boronic acid, 2-chloro-4-methylphenyl, pubchem1778, 2-chloro-4-methyl-phenyl boronic acid, acmc-209cuq, ksc524i9h, 2-chloro-p-tolylboronic acid, 2-chloro-4-methyl phenyl boronic acid
InChI Key UKYCKUPXBPLXBA-UHFFFAOYSA-N
SMILES B(C1=C(C=C(C=C1)C)Cl)(O)O
IUPAC Name (2-chloro-4-methylphenyl)boronic acid
Molecular Weight (g/mol) 170.399
PubChem CID 2773334
Formula Weight 170.40
Show More Show Less