missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-methoxyphenol, 97%

Catalog No. AAA1665214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1665214 25 g
AAA1665206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1665214 Supplier Thermo Scientific Chemicals Supplier No. A1665214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 18113-03-6 | C7H7ClO2 | 158.581 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18113-03-6
Molecular Formula C7H7ClO2
Molecular Weight (g/mol) 158.581
MDL Number MFCD00070773
InChI Key GNVRRKLFFYSLGT-UHFFFAOYSA-N
PubChem CID 87459
IUPAC Name 2-chloro-4-methoxyphenol
SMILES COC1=CC(=C(C=C1)O)Cl

Specifications

Melting Point 39°C to 44°C
Density 1.153
Flash Point 85°C (185°F)
Odor Phenol-like
Quantity 25 g
Beilstein 1936412
Synonym 3-chloro-4-hydroxyanisole, phenol, chloro-4-methoxy, phenol, 2-chloro-4-methoxy, pubchem3663, 2-chloro-4-methoxy-phenol, acmc-1c55n, phenol,2-chloro-4-methoxy, ksc445k0p, 2-chloro-4-methoxyphenol
Formula Weight 158.59
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-methoxyphenol
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 242-007-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only