missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-hydroxybenzoic Acid 98.0+%, TCI America™

Catalog No. C23495G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C23495G 5 g
C23491G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C23495G Supplier TCI America Supplier No. C23495G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 56363-84-9
Molecular Formula C7H5ClO3
Molecular Weight (g/mol) 172.56
MDL Number MFCD00798116
InChI Key WIPYZRZPNMUSER-UHFFFAOYSA-N
Synonym benzoic acid, 2-chloro-4-hydroxy, 2-chloro-4-hydroxy-benzoic acid, 2-chloro-4-hyroxybenzoic acid, pubchem4561, acmc-1b14n, ksc497o6l, 4-hydroxy-2-chlorobenzoic acid, wipyzrzpnmuser-uhfffaoysa
PubChem CID 91821
IUPAC Name 2-chloro-4-hydroxybenzoic acid
SMILES OC(=O)C1=CC=C(O)C=C1Cl

Specifications

Melting Point 159°C
Quantity 5 g
Formula Weight 172.56
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-4-hydroxybenzoic Acid