missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-chloro-4-hydroxybenzoic acid hydrate, 98%, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| CAS Min % | 97.5 |
| CAS Max % | 100.0 |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98% |
| Packaging | Glass bottle |
| Quantity | 1g |
| Formula Weight | 172.57 |
| Percent Purity | 98% |
| Chemical Name or Material | 2-chloro-4-hydroxybenzoic acid hydrate |
Safety and Handling
| EINECSNumber | 260-132-1 |
RUO – Research Use Only