missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-fluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. C27725G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C27725G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C27725G Supplier TCI America Supplier No. C27725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material 2-Chloro-4-fluorophenylboronic Acid (contains varying amounts of Anhydride)
CAS 313545-72-1
Melting Point 237°C
Color White
Molecular Formula C6H5BClFO2
Quantity 5 g
MDL Number MFCD02684295
Synonym 2-chloro-4-fluorobenzeneboronic acid, 2-chloro-4-fluorophenyl boronic acid, 2-chloro-4-fluorophenyl-boronic acid, 2-chloro-4-fluoro-phenyl boronic acid, boronic acid, 2-chloro-4-fluorophenyl, pubchem5050, acmc-209hlz, 2-chloro-4-trifluoromethylphenylboronicacid, ksc222e3l, ablock ab-12-5093
InChI Key XOFNMNLYGPKKOV-UHFFFAOYSA-N
SMILES OB(O)C1=CC=C(F)C=C1Cl
IUPAC Name (2-chloro-4-fluorophenyl)boronic acid
Molecular Weight (g/mol) 174.36
PubChem CID 17750229
Formula Weight 174.36
Physical Form Crystalline Powder
Show More Show Less