missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-fluorobenzyl Bromide 98.0+%, TCI America™

Catalog No. C20985G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C20985G 5 g
C209825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C20985G Supplier TCI America Supplier No. C20985G

Chemical Identifiers

CAS 45767-66-6
Molecular Formula C7H5BrClF
Molecular Weight (g/mol) 223.469
MDL Number MFCD00236025
InChI Key GAUUDQVOPUKGJD-UHFFFAOYSA-N
Synonym 2-chloro-4-fluorobenzyl bromide, 1-bromomethyl-2-chloro-4-fluorobenzene, 2-chloro-4-fluorobenzylbromide, alpha-bromo-2-chloro-4-fluorotoluene, benzene, 1-bromomethyl-2-chloro-4-fluoro, 2-choro-4-fluorobenzylbromide, pubchem4893, 1-bromomethyl-2-chloro-4-fluoro-benzene, ksc493m8r, 2-chloro-4-flurobenzyl bromide
PubChem CID 2725062
IUPAC Name 1-(bromomethyl)-2-chloro-4-fluorobenzene
SMILES C1=CC(=C(C=C1F)Cl)CBr

Specifications

Melting Point 39°C
Quantity 5 g
UN Number 3261
Formula Weight 223.47
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-4-fluorobenzyl Bromide
TSCA No
Recommended Storage Refrigerator