missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-fluorobenzyl bromide, 97%

Catalog No. AAB2356514
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2356514 25 g
AAB2356506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2356514 Supplier Thermo Scientific Chemicals Supplier No. B2356514

CAS: 45767-66-6 | C7H5BrClF | 223.469 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 45767-66-6
Molecular Formula C7H5BrClF
Molecular Weight (g/mol) 223.469
MDL Number MFCD00236025
InChI Key GAUUDQVOPUKGJD-UHFFFAOYSA-N
Synonym 2-chloro-4-fluorobenzyl bromide, 1-bromomethyl-2-chloro-4-fluorobenzene, 2-chloro-4-fluorobenzylbromide, alpha-bromo-2-chloro-4-fluorotoluene, benzene, 1-bromomethyl-2-chloro-4-fluoro, 2-choro-4-fluorobenzylbromide, pubchem4893, 1-bromomethyl-2-chloro-4-fluoro-benzene, ksc493m8r, 2-chloro-4-flurobenzyl bromide
PubChem CID 2725062
IUPAC Name 1-(bromomethyl)-2-chloro-4-fluorobenzene
SMILES C1=CC(=C(C=C1F)Cl)CBr

Specifications

Melting Point 35°C to 39°C
Quantity 25 g
UN Number UN3261
Beilstein 3539265
Formula Weight 223.48
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-fluorobenzyl bromide
Hazard Category H302-H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only