missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4,6-dimethyl-3-pyridinecarbonitrile 98.0+%, TCI America™

Catalog No. C26685G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C26685G 5 g
C266825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C26685G Supplier TCI America Supplier No. C26685G
Only null left

Chemical Identifiers

CAS 14237-71-9
Molecular Formula C8H7ClN2
Molecular Weight (g/mol) 166.61
MDL Number MFCD00051676
InChI Key RETJKTAVEQPNMH-UHFFFAOYSA-N
Synonym 2-chloro-4,6-dimethylnicotinonitrile, 2-chloro-3-cyano-4,6-dimethylpyridine, 2-chloro-4,6-dimethyl-nicotinonitrile, 2-chloro-4,6-dimethyl-3-pyridinecarbonitrile, 3-pyridinecarbonitrile, 2-chloro-4,6-dimethyl, acmc-1azio, chlorodimethylnicotinonitrile, ksc496m6f, 2-chloro-4,6-dimethyinicotinonitrile, 4,6-dimethyl-2-chloro-nicotinonitrile
PubChem CID 223261
IUPAC Name 2-chloro-4,6-dimethylpyridine-3-carbonitrile
SMILES CC1=CC(C)=C(C#N)C(Cl)=N1

Specifications

Melting Point 99°C
Color White-Yellow
Quantity 5 g
UN Number 3439
Formula Weight 166.61
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-4,6-dimethyl-3-pyridinecarbonitrile