missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-3-iodopyridine 98,0+%, TCI America™
Spécifications
Spécifications
| Melting Point | 95°C |
| Quantity | 1 g |
| Formula Weight | 239.44 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Poudre cristalline |
| Chemical Name or Material | 2-Chloro-3-iodopyridine |
Sécurité et manipulation
| TSCA | No |