missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-fluoropyridine 98.0+%, TCI America™

Catalog No. C24475G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C24475G 5 g
C244725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C24475G Supplier TCI America Supplier No. C24475G

Chemical Identifiers

CAS 17282-04-1
Molecular Formula C5H3ClFN
Molecular Weight (g/mol) 131.534
MDL Number MFCD03095302
InChI Key SVAZIMBLBHOVIR-UHFFFAOYSA-N
Synonym 3-fluoro-2-chloropyridine, 2-chloro-3-fluoro pyridine, pyridine, 2-chloro-3-fluoro, 2-chloro-3-fluoro-pyridine, pubchem2532, acmc-1cc0d, ksc174s3h, 2-chloro-3-fluoropyridine, 2-chloro-3-fluoropyridine 1g
PubChem CID 2782791
IUPAC Name 2-chloro-3-fluoropyridine
SMILES C1=CC(=C(N=C1)Cl)F

Specifications

Color Yellow
Boiling Point 80°C
Quantity 5 g
Formula Weight 131.53
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloro-3-fluoropyridine