missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-fluorophenylboronic acid, 97%

Catalog No. AC430370010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC430370010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC430370010 Supplier Thermo Scientific Chemicals Supplier No. 430370010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 871329-52-1 | C6H5BClFO2 | 174.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 871329-52-1
Molecular Formula C6H5BClFO2
Molecular Weight (g/mol) 174.36
MDL Number MFCD07363741
InChI Key TYOGIUGNZOBBHE-UHFFFAOYSA-N
Synonym 2-chloro-3-fluorophenyl boronic acid, 1-borono-2-chloro-3-fluorobenzene, 2-chloro-3-fluorobenzeneboronic acid, pubchem19756, acmc-209qfx, 2-chloro-3-fluoro-phenylboronic acid, 2-chloro-3-fluoro-benzeneboronic acid, 2-chloro-3-fluoro-phenyl boronic acid, 2-chloro-3-fluorophenyl ;boronic acid
PubChem CID 22309459
IUPAC Name (2-chloro-3-fluorophenyl)boronic acid
SMILES OB(O)C1=C(Cl)C(F)=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 115.8°C to 115.9°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 174.37
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-chloro-3-fluorophenylboronic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only