missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-1-methylimidazole-5-boronic acid pinacol ester, 95%

Catalog No. AAH31692MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH31692MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH31692MD Supplier Thermo Scientific Chemicals Supplier No. H31692MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1315281-29-8 | C10H16BClN2O2 | 242.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Sparingly soluble in water (0.23 g/L)

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 1315281-29-8
Molecular Formula C10H16BClN2O2
Molecular Weight (g/mol) 242.51
MDL Number MFCD11100959
InChI Key MOQXZCYTVMKINQ-UHFFFAOYSA-N
Synonym 2-chloro-1-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-imidazole, 2-chloro-1-methylimidazole-5-boronic acid pinacol ester, 2-chloro-1-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl imidazole, 2-chloro-1-methyl-1h-imidazole-5-boronic acid pinacol ester, 2-chloro-1-methyl-1h-imidazole-5-boronic acid, pinacol ester, 2-chloro-1-methyl-1h-imidazol-5-yl boronic acid pinacol ester, 2-chloro-1-methyl-5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-imidazole, 2-chloro-1-methyl-5-tetramethyl-1,3,2-dioxaborolan-2-yl imidazole
PubChem CID 45588157
IUPAC Name 2-chloro-1-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)imidazole
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CN=C(N2C)Cl

Specifications

Melting Point 131°C to 135°C
Quantity 250 mg
Solubility Information Sparingly soluble in water (0.23g/L)
Formula Weight 242.52
Percent Purity 95%
Chemical Name or Material 2-Chloro-1-methylimidazole-5-boronic acid pinacol ester
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only