missing translation for 'onlineSavingsMsg'
Learn More

(2-Carboxyethyl)phenylphosphinic Acid 98.0+%, TCI America™

Catalog No. C28875G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C28875G 5 g
C288725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C28875G Supplier TCI America Supplier No. C28875G

Chemical Identifiers

CAS 14657-64-8
Molecular Formula C9H11O4P
Molecular Weight (g/mol) 214.157
MDL Number MFCD08704562
InChI Key MORLYCDUFHDZKO-UHFFFAOYSA-N
PubChem CID 11287340
IUPAC Name 3-[hydroxy(phenyl)phosphoryl]propanoic acid
SMILES C1=CC=C(C=C1)P(=O)(CCC(=O)O)O

Specifications

Melting Point 160°C
Color White
Quantity 5 g
Formula Weight 214.16
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (2-Carboxyethyl)phenylphosphinic Acid