missing translation for 'onlineSavingsMsg'
Learn More

2-Carboxybenzaldehyde, 99%, Thermo Scientific™

Numéro de catalogue. p-3757193
Modifier l'affichage
Click to view available options
Quantité:
25g
100g
Emballage:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
AC152080250 25g Glass bottle
AC152081000 100g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AC152080250 Fournisseur Thermo Scientific Chemicals Code fournisseur 152080250

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Identifiants chimiques

CAS 119-67-5
Formule moléculaire C8H6O3
Poids moléculaire (g/mol) 150.13
Numéro MDL MFCD00003336
Clé InChI DYNFCHNNOHNJFG-UHFFFAOYSA-N
Synonyme 2-carboxybenzaldehyde, phthalaldehydic acid, o-formylbenzoic acid, benzoic acid, 2-formyl, o-carboxybenzaldehyde, benzoic acid, formyl, benzaldehyde-2-carboxylic acid, formylbenzoic acid, carboxybenzaldehyde, pubchem7365
PubChem CID 8406
ChEBI CHEBI:17605
Nom de l’IUPAC 2-formylbenzoic acid
SOURIRES C1=CC=C(C(=C1)C=O)C(=O)O

Spécifications

Nom ou substance chimique 2-Carboxybenzaldehyde
CAS % minimum 98.5
CAS % maximal 100.0
Spectre infrarouge Authentic
Fourchette de pourcentages d’essai 98.5% min. (HPLC)
Emballage Glass bottle
Formule linéaire HO2CC6H4CHO
Quantité 25g
Beilstein 10, 666
Renseignements sur la solubilité Solubility in water: soluble
Poids de formule 150.13
Pourcentage de pureté 99%
Afficher plus de résultats Afficher moins de résultats
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 204-342-3

RUO – Research Use Only