missing translation for 'onlineSavingsMsg'
Learn More

2-Butyne-1,4-diol, 99%

Catalog No. AC108082500
Change view
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC108082500 250 g Plastic bottle
AC108080010 1 kg Plastic bottle
AC108080025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC108082500 Supplier Thermo Scientific Chemicals Supplier No. 108082500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 110-65-6 | C4H6O2 | 86.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 110-65-6
Molecular Formula C4H6O2
Molecular Weight (g/mol) 86.09
MDL Number MFCD00002915
InChI Key DLDJFQGPPSQZKI-UHFFFAOYSA-N
Synonym 2-butyne-1,4-diol, butynediol, 1,4-dihydroxy-2-butyne, 1,4-butynediol, bis hydroxymethyl acetylene, 2-butynediol, 2-butin-1,4-diol, agrisynth b3d, unii-axh202fpqm, 1,4-butynediol van
PubChem CID 8066
ChEBI CHEBI:16413
IUPAC Name but-2-yne-1,4-diol
SMILES C(C#CCO)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 54.0°C to 58.0°C
Color Yellow
Density 1.2000g/mL
Boiling Point 238.0°C
Flash Point 152°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Refractive Index 1.4804
Linear Formula HOCH2C≡CCH2OH
Quantity 250 g
Beilstein 01, I, 261
Specific Gravity 1.2
Solubility Information Solubility in water: 2960g/L (20°C). Other solubilities: soluble in aqueous acids,soluble in alcohol and acetone,insoluble in ether and benzene,soluble in polar solvents
Formula Weight 86.09
Percent Purity 99%
Physical Form Crystalline Platelets or Flakes
Chemical Name or Material 2-Butyne-1, 4-diol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause an allergic skin reaction.
Toxic if inhaled.
Causes severe skin burns and eye damage.
May cause damage to organs through prolonged or repeated exposure.
Toxic if swallowed.
Toxic in contact w
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 203-788-6
Recommended Storage Product darkens on storage

RUO – Research Use Only