missing translation for 'onlineSavingsMsg'
Learn More

2-Butyl-5-chloro-1H-imidazole-4-carboxaldehyde 98.0+%, TCI America™

Catalog No. B291425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B291425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B291425G Supplier TCI America Supplier No. B291425G

Chemical Identifiers

CAS 83857-96-9
Molecular Formula C8H11ClN2O
Molecular Weight (g/mol) 186.64
MDL Number MFCD01934396
InChI Key JLVIHQCWASNXCK-UHFFFAOYSA-N
Synonym 2-butyl-4-chloro-5-formylimidazole, 2-butyl-4-chloro-1h-imidazole-5-carbaldehyde, unii-7h170u0swe, 2-butyl-5-chloroimidazole-4-carbaldehyde, 2-butyl-5-chloro-3h-imidazole-4-carbaldehyde, 2-butyl-4 5-chloro-5 4 imidazolecarboxaldehyde, 1h-imidazole-4-carboxaldehyde, 2-butyl-5-chloro, 2-butyl-4-chloro-1h-imidazole-5-carboxaldehyde, 2-butyl-4-chloro-5-formylimidazloe, 2-n-butyl-4-chloro-5-formylimidazole
PubChem CID 55176
IUPAC Name 2-butyl-5-chloro-1H-imidazole-4-carbaldehyde
SMILES CCCCC1=NC(C=O)=C(Cl)N1

Specifications

Melting Point 99°C
Quantity 25 g
UN Number 3077
Formula Weight 186.64
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Butyl-5-chloro-1H-imidazole-4-carboxaldehyde