missing translation for 'onlineSavingsMsg'
Learn More

2-Butoxyethyl Acetate, Spectrum™ Chemical

Catalog No. 18608206
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608206 4 L Amber Glass Bottle
18608204 1 L Amber Glass Bottle
18608205 20 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18608206 Supplier Spectrum Chemical Mfg Cor Supplier No. B11814LTGL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 112-07-2
Molecular Formula C8H16O3
Molecular Weight (g/mol) 160.21
InChI Key NQBXSWAWVZHKBZ-UHFFFAOYSA-N
IUPAC Name 2-butoxyethyl acetate
SMILES CCCCOCCOC(C)=O

Specifications

Acidity 0.0002
CAS Max % 1
Boiling Point 192°C (378°F)
Packaging Amber Glass Bottle
Quantity 4 L
Specific Gravity 0.940 - 0.944
Grade Reagent
Physical Form Liquid
Chemical Name or Material 2-Butoxyethyl Acetate

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.