missing translation for 'onlineSavingsMsg'
Learn More

2-Butoxyethanol, 97%, Spectrum™ Chemical

Catalog No. 18608203
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608203 4 L Amber Glass Bottle
18608200 1 L Amber Glass Bottle
18608201 20 L Amber Glass Bottle
18608202 200 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18608203 Supplier Spectrum Chemical Mfg Cor Supplier No. B11774LTGL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 111-76-2

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 4 L
Molecular Weight (g/mol) 118.18
Percent Purity 0.97
Grade Reagent
Physical Form Liquid
Chemical Name or Material 2-Butoxyethanol

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.