missing translation for 'onlineSavingsMsg'
Learn More

2-Butanone, puriss. p.a., ACS Reagent, Reag. Ph. Eur., ≥99.5% (GC), Solstice

Catalog No. 60046236
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60046236 4 x 2.5 L Glass Bottle
60046232 1 L Glass Bottle
60046233 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60046236 Supplier Honeywell Inc Supplier No. 334074X2.5L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Puriss. p.a., ACS Reagent, Reag. Ph. Eur.,≥99.5% (GC)

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications

Specifications

Melting Point -87°C
Density 0.805g/mL at 25°C
Quantity 4 x 2.5 L
Flash Point −3°C (26.6°F)
Odor Solvent like
Packaging Glass Bottle
Boiling Point 79°C to 81°C
Solubility Information 290g/L at 20°C