missing translation for 'onlineSavingsMsg'
Learn More

2-Butanol 99.0+%, TCI America™

Catalog No. B070525ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B070525ML 25 mL
B0705500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B070525ML Supplier TCI America Supplier No. B070525ML

Chemical Identifiers

CAS 78-92-2
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00004569
InChI Key BTANRVKWQNVYAZ-UHFFFAOYNA-N
Synonym 2-butanol, sec-butanol, sec-butyl alcohol, 2-hydroxybutane, 2-butyl alcohol, s-butyl alcohol, butylene hydrate, s-butanol, 1-methyl-1-propanol, 1-methylpropyl alcohol
PubChem CID 6568
ChEBI CHEBI:35687
IUPAC Name butan-2-ol
SMILES CCC(C)O

Specifications

Melting Point -115°C
Color Colorless
Boiling Point 100°C
Quantity 25 mL
UN Number 1120
Formula Weight 74.12
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Butanol
EINECSNumber (2)-3049
RTECSNumber EO1750000
TSCA Yes