missing translation for 'onlineSavingsMsg'
Learn More

2-Bromostyrene (stabilized with TBC) 95.0+%, TCI America™

Catalog No. B23555G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B23555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B23555G Supplier TCI America Supplier No. B23555G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2039-88-5
Molecular Formula C8H7Br
Molecular Weight (g/mol) 183.048
MDL Number MFCD00000076
InChI Key SSZOCHFYWWVSAI-UHFFFAOYSA-N
Synonym 2-bromostyrene, 1-bromo-2-vinylbenzene, o-bromostyrene, benzene, 1-bromo-2-ethenyl, unii-9a96gp6z5d, styrene, o-bromo, 2-bromo-styrene, o-bromovinylbenzene, pubchem23975, 2-bromo-1-vinylbenzene
PubChem CID 16264
IUPAC Name 1-bromo-2-ethenylbenzene
SMILES C=CC1=CC=CC=C1Br

Specifications

Melting Point -53°C
Color Yellow
Boiling Point 61°C
Quantity 5 g
Formula Weight 183.05
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromostyrene (stabilized with TBC)
TSCA No
Recommended Storage Refrigerator