missing translation for 'onlineSavingsMsg'
Learn More

2-Bromostyrene, 96%, stab. with ca 0.05% 4-tert-butylcatechol

Catalog No. AAB2418106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2418106 5 g
AAB2418114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2418106 Supplier Thermo Scientific Chemicals Supplier No. B2418106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2039-88-5 | C8H7Br | 183.048 g/mol

2-Bromostyrene is used as a cross linking agent in fire-resistant bromostyrene-crosslinked polyesters. Further, it is used to prepare trans-1-(2-bromophenyl)-2-(3-bromophenyl)ethene by reacting with 1-bromo-3-iodo-benzene, involving palladium(II) acetate as catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromostyrene is used as a cross linking agent in fire-resistant bromostyrene-crosslinked polyesters. Further, it is used to prepare trans-1-(2-bromophenyl)-2-(3-bromophenyl)ethene by reacting with 1-bromo-3-iodo-benzene, involving palladium(II) acetate as catalyst.

Solubility
Miscible with methanol.

Notes
Incompatible with acids, bases, oxidizing agents and halogens.

Chemical Identifiers

CAS 2039-88-5
Molecular Formula C8H7Br
Molecular Weight (g/mol) 183.048
MDL Number MFCD00000076
InChI Key SSZOCHFYWWVSAI-UHFFFAOYSA-N
Synonym 2-bromostyrene, 1-bromo-2-vinylbenzene, o-bromostyrene, benzene, 1-bromo-2-ethenyl, unii-9a96gp6z5d, styrene, o-bromo, 2-bromo-styrene, o-bromovinylbenzene, pubchem23975, 2-bromo-1-vinylbenzene
PubChem CID 16264
IUPAC Name 1-bromo-2-ethenylbenzene
SMILES C=CC1=CC=CC=C1Br

Specifications

Melting Point -53°C
Density 1.416
Boiling Point 102°C to 104°C (22 mmHg)
Flash Point 85°C (185°F)
Refractive Index 1.592
Quantity 5 g
Beilstein 2323537
Solubility Information Miscible with methanol.
Formula Weight 183.05
Percent Purity 96%
Chemical Name or Material 2-Bromostyrene, stabilized with ≈0.05% 4-tert-butylcatechol
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 218-027-3
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only