missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoquinoline, 96%

Catalog No. AAH2730214
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2730214 25 g
AAH2730203 1 g
AAH2730206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH2730214 Supplier Thermo Scientific Chemicals Supplier No. H2730214
Only null left

CAS: 2005-43-8 | C9H6BrN | 208.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2005-43-8
Molecular Formula C9H6BrN
Molecular Weight (g/mol) 208.06
MDL Number MFCD00234480
InChI Key QKJAZPHKNWSXDF-UHFFFAOYSA-N
Synonym quinoline, 2-bromo, bromoquinoline, pubchem21827, acmc-1cfcf, 2-bromoquinoline, ksc201o8j, 2-bromoquinoline 1g, abbypharma ap-12-10890
PubChem CID 2762756
IUPAC Name 2-bromoquinoline
SMILES BrC1=CC=C2C=CC=CC2=N1

Specifications

Quantity 25 g
Formula Weight 208.06
Percent Purity 96%
Chemical Name or Material 2-Bromoquinoline
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only