missing translation for 'onlineSavingsMsg'
Learn More

2-Bromopyridine-5-boronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. B33071G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B33071G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B33071G Supplier TCI America Supplier No. B33071G

Specifications

Chemical Name or Material 2-Bromopyridine-5-boronic Acid (contains varying amounts of Anhydride)
CAS 223463-14-7
Melting Point 165°C
Molecular Formula C5H5BBrNO2
Quantity 1 g
MDL Number MFCD03411558
Synonym 2-bromopyridine-5-boronic acid, 6-bromopyridin-3-yl boronic acid, 6-bromopyridine-3-boronic acid, 2-bromo-5-pyridineboronic acid, 6-bromopyridin-3-lboronic acid, 2-bromo-5-pyridylboronic acid, 2-bromopyridyl-5-boronic acid, 6-bromo-3-pyridylboronic acid, 6-bromopyridin-3-yl-3-boronic acid, 6-bromo-3-pyridinylboronic acid
InChI Key BCYWDUVHAPHGIP-UHFFFAOYSA-N
SMILES OB(O)C1=CN=C(Br)C=C1
IUPAC Name (6-bromopyridin-3-yl)boronic acid
Molecular Weight (g/mol) 201.81
PubChem CID 2762706
Formula Weight 201.81
Physical Form Crystalline Powder
Show More Show Less