missing translation for 'onlineSavingsMsg'
Learn More

2-Bromopropionyl Bromide 98.0+%, TCI America™

Catalog No. B0649500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0649500G 500 g
B064925G 25 g
B0649100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B0649500G Supplier TCI America Supplier No. B0649500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 563-76-8
Molecular Formula C3H4Br2O
Molecular Weight (g/mol) 215.872
MDL Number MFCD00000142
InChI Key ILLHORFDXDLILE-UHFFFAOYSA-N
Synonym 2-bromopropionyl bromide, propanoyl bromide, 2-bromo, 2-bromopropionylbromide, 2-bromo propionyl bromide, 2-bromopropionic acid bromide, .alpha.-bromopropionyl bromide, 2-bromopropanyl bromide, a-bromopropionyl bromide, 2-bromo-propionyl bromide, alpha-bromopropionylbromide
PubChem CID 79047
IUPAC Name 2-bromopropanoyl bromide
SMILES CC(C(=O)Br)Br

Specifications

Color Yellow
Boiling Point 155°C
Quantity 500 g
UN Number 3265
Formula Weight 215.87
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 2-Bromopropionyl Bromide
EINECSNumber (2)-1168
TSCA Yes