missing translation for 'onlineSavingsMsg'
Learn More

2-Bromopropionic Acid 98.0+%, TCI America™

Catalog No. B0644500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0644500G 500 g
B064425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0644500G Supplier TCI America Supplier No. B0644500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 598-72-1
Molecular Formula C3H5BrO2
Molecular Weight (g/mol) 152.98
MDL Number MFCD00004211,MFCD00145205
InChI Key MONMFXREYOKQTI-UHFFFAOYNA-N
Synonym 2-bromopropionic acid, propanoic acid, 2-bromo, dl-2-bromopropionic, propionic acid, 2-bromo, alpha-bromopropionic acid, dl-2-bromopropionic acid, 2-bromo-propionic acid, .alpha.-bromopropionic acid, propanoic acid, bromo, propanoicacid, 2-bromo
PubChem CID 11729
IUPAC Name 2-bromopropanoic acid
SMILES CC(Br)C(O)=O

Specifications

Color Yellow
Boiling Point 205°C
Quantity 500 g
UN Number 3265
Formula Weight 152.98
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Bromopropionic Acid
EINECSNumber (9)-1167
RTECSNumber UA2451715
TSCA Yes