missing translation for 'onlineSavingsMsg'
Learn More

2-Bromopropene, 99%, stabilized

Catalog No. AC107340500
Change view
Click to view available options
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC107340500 50 mL Glass bottle
AC107340100 10 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC107340500 Supplier Thermo Scientific Chemicals Supplier No. 107340500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 557-93-7 | C3H5Br | 120.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 557-93-7
Molecular Formula C3H5Br
Molecular Weight (g/mol) 120.98
MDL Number MFCD00000140
InChI Key PHMRPWPDDRGGGF-UHFFFAOYSA-N
Synonym 2-bromopropene, isopropenyl bromide, 1-propene, 2-bromo, 2-bromo-1-propene, 2-bromopropylene, propene, 2-bromo, isopropylene bromide, alpha-methylvinyl bromide, .alpha.-methylvinyl bromide, 2-bromo-propene
PubChem CID 11202
IUPAC Name 2-bromoprop-1-ene
SMILES CC(=C)Br

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.3600g/mL
Boiling Point 47.0°C to 49.0°C
Flash Point 4°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3C(Br)=CH2
Refractive Index 1.4430 to 1.4450
Quantity 50 mL
Beilstein 01,2
Specific Gravity 1.36
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
Formula Weight 120.98
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Bromopropene, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 209-185-4
RTECSNumber UC7085000
TSCA TSCA

RUO – Research Use Only