missing translation for 'onlineSavingsMsg'
Learn More

2-Bromophenanthrene 97.0+%, TCI America™

Numéro de catalogue. B4671200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
B4671200MG 200 mg
B46711G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. B4671200MG Fournisseur TCI America Code fournisseur B4671200MG
Il en reste null

Identifiants chimiques

CAS 62162-97-4
Molecular Formula C14H9Br
Molecular Weight (g/mol) 257.13
InChI Key SQTPFYJEKHTINP-UHFFFAOYSA-N
PubChem CID 12491433
IUPAC Name 2-bromophenanthrene
SMILES C1=CC=C2C(=C1)C=CC3=C2C=CC(=C3)Br

Spécifications

Melting Point 97°C
Color White-Yellow
Boiling Point 185°C
Quantity 200 mg
Formula Weight 257.13
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromophenanthrene